missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C. botulinum BoNT-B Light Chain Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marke: R&D Systems MAB5420
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
BoNT-B Light Chain Monoclonal specifically detects BoNT-B Light Chain in C. botulinum samples. It is validated for Western Blot.
Spezifikation
| BoNT-B Light Chain | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P10844 | |
| boNT/B | |
| E. coli-derived recombinant Clostridium BoNT-B Light Chain Pro2-His428 Accession # P10844 | |
| 100 μg | |
| Primary | |
| Detects C. botulinum BoNT-B Light Chain in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant Light Chains of BoNT-A, -C1, -D, -E, -F, or -G is observed. | |
| C. botulinum | |
| Purified |
| Western Blot | |
| 579631 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BoNTB Light Chain, DcD | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6030102 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur