missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C. botulinum BoNT-B Light Chain Antibody, R&D Systems™

Sheep Polyclonal Antibody
196.00 € - 491.00 €
Spezifikation
| Antigen | BoNT-B Light Chain |
|---|---|
| Konzentration | LYOPH |
| Verdünnung | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Anwendungen | Western Blot, Immunoprecipitation |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18759378
![]() |
R&D Systems
AF5420-SP |
25 μg |
196.00 €
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18099031
|
R&D Systems
AF5420 |
100 μg |
491.00 €
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
BoNT-B Light Chain Polyclonal specifically detects BoNT-B Light Chain in C. botulinum samples. It is validated for Western Blot, Immunoprecipitation.Spezifikation
| BoNT-B Light Chain | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Sheep | |
| C. botulinum | |
| BoNTB Light Chain, DcD | |
| boNT/B | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| P10844 | |
| 6030102 | |
| E. coli-derived recombinant Clostridium BoNT-B Light Chain Pro2-His428 Accession # P10844 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects C. botulinum BoNT-B Light Chain in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant Light Chains of BoNT-A, -E, or recombinant Heavy Chains of BoNT-C, or -D is observed. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts