missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Met inhibitor 1, MedChemExpress
Beschreibung
c-Met inhibitor 1 is an inhibitor of the c-Met receptor signaling pathway useful for the treatment of cancer including gastric, glioblastoma, and pancreatic cancer.
Spezifikation
Spezifikation
| Chemischer Name oder Material | c-Met inhibitor 1 |
| Physikalische Form | Solid |
| CAS | 1357072-61-7 |
| Farbe | Off-White |
| Menge | 100 mg |
| Hinweise zur Reinheitsqualität | Research |
| Summenformel | C17H14N8S |
| Zur Verwendung mit (Anwendung) | Cancer-Kinase/protease |
| Löslichkeitsinformationen | DMSO : 20 mg/mL (55.19 mM; Need ultrasonic) |
| SMILES | CN1N=CC(C2=NN3C(C=C2)=NN=C3SC4=CC5=CN(C)N=C5C=C4)=C1 |
| Mehr anzeigen |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?