Learn More
C12orf41 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558416
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: FSDDLDVVGD GMQCPPSPLL FDPSLTLEDH LVKEIAEDPV DILGQMQMAG DGCRSQGSRN SEKASAPLSQ SGLATANGKP EP Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 76%.
C12orf41 Polyclonal antibody specifically detects C12orf41 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
C12orf41 | |
Polyclonal | |
Unconjugated | |
KANSL2 | |
2310037I24Rik; B930009D17; C12orf41; Kansl2; KAT8 regulatory NSL complex subunit 2; non-specific lethal 2 homolog; NSL complex protein NSL2; NSL2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54934 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H9L4 | |
KANSL2 | |
Recombinant protein corresponding to Human C12orf41 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only