Learn More
C14orf80 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558637
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PAPHMEAEGP VDVRHVQWLM GKLRFRWRQL VSSQQEQCAL LSKIHLYTRG CHSDQSLSHL SVTEAEMLRD PEGGQQVSGA GAAQNLDLA Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 62%.
C14orf80 may play a role in counteracting perturbation of actin filaments, such as after treatment with the actin depolymerizing microbial metabolite Chivosazole F.Spezifikation
C14orf80 | |
Polyclonal | |
Unconjugated | |
TEDC1 | |
C14orf80; chromosome 14 open reading frame 80; TEDC1; Tubulin epsilon and delta complex protein 1; uncharacterized protein C14orf80 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
283643 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86SX3 | |
TEDC1 | |
Recombinant protein corresponding to Human C14orf80 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only