missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C15orf48 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5122186
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This gene was first identified in a study of human esophageal squamous cell carcinoma tissues. Levels of both the message and protein are reduced in carcinoma samples. In adult human tissues, this gene is expressed in the esophagus, stomach, small intestine, colon and placenta. Alternatively spliced transcript variants that encode the same protein have been identified.
Spezifikation
| C15orf48 | |
| Polyclonal | |
| Unconjugated | |
| C15orf48 | |
| C15orf48; chromosome 15 open reading frame 48; FOAP-11; NMES1; normal mucosa of esophagus specific 1; normal mucosa of esophagus-specific gene 1 protein; Protein FOAP-11 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 84419 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9C002 | |
| C15orf48 | |
| Met1-Lys83 (Accession Q9C002), with N-terminal His and GST Tag. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur