missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C15orf48 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
208.00 € - 453.00 €
Spezifikation
| Antigen | C15orf48 |
|---|---|
| Konzentration | 0.5 mg/mL |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
17753584
|
Invitrogen™
PA5122187 |
20 μL |
208.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
17743584
![]() |
Invitrogen™
PA5122186 |
100 μL |
453.00 €
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
This gene was first identified in a study of human esophageal squamous cell carcinoma tissues. Levels of both the message and protein are reduced in carcinoma samples. In adult human tissues, this gene is expressed in the esophagus, stomach, small intestine, colon and placenta. Alternatively spliced transcript variants that encode the same protein have been identified.Spezifikation
| C15orf48 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9C002 | |
| 84419 | |
| Met1-Lys83 (Accession Q9C002), with N-terminal His and GST Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| C15orf48 |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| C15orf48; chromosome 15 open reading frame 48; FOAP-11; NMES1; normal mucosa of esophagus specific 1; normal mucosa of esophagus-specific gene 1 protein; Protein FOAP-11 | |
| C15orf48 | |
| IgG | |
| Protein A, Antigen affinity chromatography | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts