Learn More
C15orf57 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559676
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GSGLRFQMKM FESADSTATR SGQDLWAEIC SCLPNPEQED GANNAFSDSF VDSCPEGEGQ REVADFAVQP AVKPWAPLQD SEVYLASLEK KL Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 74%.
C15orf57 is a protein coding gene and its exact function is unknown.Spezifikation
C15orf57 | |
Polyclonal | |
Unconjugated | |
CCDC32 | |
C15orf57; CCDC32; chromosome 15 open reading frame 57; coiled-coil domain containing 32; Coiled-coil domain-containing protein 32; uncharacterized protein C15orf57 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90416 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.7 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BV29 | |
CCDC32 | |
Recombinant protein corresponding to Human C15orf57 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only