Learn More
C17orf58 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554672
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: FRVHMLALDS SSCNKPCPEF KPGSRYIVMG HIYHKRRQLP TALLQVLRGR LRPGDGLLRS SSSYVKRFNR KREGQIQGAI H Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 86%.
C17orf58 Polyclonal antibody specifically detects C17orf58 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
C17orf58 | |
Polyclonal | |
Unconjugated | |
C17orf58 | |
C17orf58; chromosome 17 open reading frame 58; UPF0450 protein C17orf58 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284018 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2M2W7 | |
C17orf58 | |
Recombinant protein corresponding to Human C17orf58 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only