Learn More
C17orf74 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554860
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LPPPSLYLSP ELRCMPKRVE ARSELRLQSY GRHGSQSRLW GNVEAEQWAS SPPPPHRLPP NPSWVPVGHS PYPSVGWMLY DSWDQ Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.
C17orf74 is a protein coding gene.Spezifikation
C17orf74 | |
Polyclonal | |
Unconjugated | |
SPEM2 | |
C17orf74; chromosome 17 open reading frame 74; SPEM2; uncharacterized protein C17orf74; Uncharacterized protein SPEM2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
201243 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q0P670 | |
SPEM2 | |
Recombinant protein corresponding to Human C17orf74 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only