Learn More
C17orf98 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562460
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: AYLSECRLRL EKGFILDGVA VSTAARAYGR SRPKLWSAIP PYNAQQDYHA RSYFQSHVVP PLLRKTDQDH GGTGRDGWI Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 72%.
C17orf98 Polyclonal antibody specifically detects C17orf98 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
C17orf98 | |
Polyclonal | |
Unconjugated | |
C17orf98 | |
C17orf98; chromosome 17 open reading frame 98; uncharacterized protein C17orf98 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
388381 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A8MV24 | |
C17orf98 | |
Recombinant protein corresponding to Human C17orf98 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only