Learn More
C19orf54 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563648
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: CGLVALWMAG TLLSPPSGVP LERLIRVATE RGYTAQGEMF SVADMGRLAQ EVLGCQAKLL SGGLGGPNRD LVL Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 79%.
The specific function of this protein remains unknown.Spezifikation
C19orf54 | |
Polyclonal | |
Unconjugated | |
C19orf54 | |
C19orf54; chromosome 19 open reading frame 54; UPF0692 protein C19orf54 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
284325 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5BKX5 | |
C19orf54 | |
Recombinant protein corresponding to Human C19orf54 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only