Learn More
C19orf81 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563824
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RNRWLIAVTD FQTRSRLLRS GLSPRGLAHQ IVRHDDLLLG DYRLHLRRSL VRRRMLEALG AEPNEEA Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 28%.
C19orf81 Polyclonal antibody specifically detects C19orf81 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
C19orf81 | |
Polyclonal | |
Unconjugated | |
C19orf81 | |
C19orf81; chromosome 19 open reading frame 81; putative uncharacterized protein C19orf81 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
342918 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
C9J6K1 | |
C19orf81 | |
Recombinant protein corresponding to Human C19orf81 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only