missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1D Rabbit anti-Human, Clone: 4, Novus Biologicals™
Rabbit Monoclonal Antibody
Marke: Novus Biologicals NBP2-90103-50UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
C1D Monoclonal specifically detects C1D in Human samples. It is validated for Western Blot.
Spezifikation
| C1D | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 10438 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 4 | |
| Western Blot 1:500-1:2000 | |
| C1D DNA-binding protein, C1D nuclear receptor corepressor, C1D nuclear receptor co-repressor, hC1D, MGC12261, MGC14659, nuclear DNA-binding protein, nuclear nucleic acid-binding protein C1D, small unique nuclear receptor corepressor, small unique nuclear receptor co-repressor, SUN-CoR, SUNCORLRP1 | |
| Recombinant Human C1D protein | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto