Learn More
C1D Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558025
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MAGEEINEDY PVEIHEYLSA FENSIGAVDE MLKTMMSVSR NELLQKLDPL EQAKVDLVSA YTLNSMF Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
The protein encoded by this gene is a DNA binding and apoptosis-inducing protein and is localized in the nucleus. It is also a Rac3-interacting protein which acts as a corepressor for the thyroid hormone receptor. This protein is thought to regulate TRAX/Translin complex formation. Alternate splicing results in multiple transcript variants that encode the same protein. Multiple pseudogenes of this gene are found on chromosome 10.Spezifikation
C1D | |
Polyclonal | |
Unconjugated | |
C1D | |
1110036E10Rik; AI875855; C1D; C1D DNA-binding protein; C1D nuclear receptor corepressor; C1D nuclear receptor co-repressor; hC1D; LRP1; mC1D; MGC12261; MGC14659; MGC188504; nuclear DNA binding protein; nuclear DNA binding protein, pseudogene 1; nuclear DNA-binding protein; nuclear nucleic acid-binding protein C1D; rCG23324; RGD1560600; Rrp47; small unique nuclear co-repressor; small unique nuclear receptor corepressor; small unique nuclear receptor co-repressor; SUNCOR; Sun-CoR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10438 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13901 | |
C1D | |
Recombinant protein corresponding to Human C1D | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only