Learn More
C1orf110 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555907
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LYKHFDLERK NAKQAEARLD QRLQRLKVIC LYHVKLLTWE QRQLQKELQR LQQETMKKKF SSYLGNGFQK RPEDVLVFSP QGR Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 71%.
C1orf110 Polyclonal antibody specifically detects C1orf110 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
C1orf110 | |
Polyclonal | |
Unconjugated | |
CCDC190 | |
1700084C01Rik; C1orf110; Ccdc190; coiled-coil domain containing 190; coiled-coil domain-containing protein 190; coiled-coil domain-containing protein C1orf110 homolog; RGD1560672; RIKEN cDNA 1700084C01 gene; similar to novel protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
339512 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86UF4 | |
CCDC190 | |
Recombinant protein corresponding to Human C1orf110 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only