Learn More
C1orf69 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA556498
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: FPVRFLDPLP TSGITPGATV LTASGQTVGK FRAGQGNVGL ALLWSEKIKG PLHIRASEGA QVALAASVPD WWPTVSK Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 65%.
Involved in the maturation of mitochondrial 4Fe-4S proteins functioning late in the iron-sulfur cluster assembly pathway.Spezifikation
C1orf69 | |
Polyclonal | |
Unconjugated | |
IBA57 | |
4930543L23Rik; A230051G13Rik; C1orf69; IBA57; IBA57 homolog, iron-sulfur cluster assembly; IBA57, iron-sulfur cluster assembly homolog; IBA57, iron-sulfur cluster assembly homolog (S. cerevisiae); iron-sulfur cluster assembly factor for biotin synthase- and aconitase-like mitochondrial proteins, with a mass of 57kDa; iron-sulfur cluster assembly factor homolog; MMDS3; putative transferase C1orf69 homolog, mitochondrial; putative transferase C1orf69, mitochondrial; putative transferase CAF17 homolog, mitochondrial; Putative transferase CAF17, mitochondrial; RGD1306000; SPG74 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
200205 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T440 | |
IBA57 | |
Recombinant protein corresponding to Human C1orf69 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only