Learn More
C21orf59 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555990
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: QVLKKTIEEA KAIISKKQVE AGVCVTMEMV KDALDQLRGA VMIVYPMGLP PYDPIRMEFE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.
The function of the C21orf59 protein remains unknown.Spezifikation
C21orf59 | |
Polyclonal | |
Unconjugated | |
C21ORF59 | |
C21orf48; C21orf59; CFAP298; chromosome 21 open reading frame 59; CILD26; Cilia- and flagella-associated protein 298; FBB18; Kur; prostate cancer upregulated protein 1; Protein kurly homolog; UPF0769 protein C21orf59 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56683 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P57076 | |
C21ORF59 | |
Recombinant protein corresponding to Human C21orf59 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Research Use Only