Learn More
C2CD4C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563206
Weitere Details : Netto-Gewicht : 0.02000kg
Description
Immunogen sequence: PSTSEQNLAS AAPRQTPRSP RLPAKLAAES KSLLKAATRH VIQIESAEDW LSEEATDADP QAQGAMSLPS VPKAQTSYGF AMLAES Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.
C2CD4C Polyclonal antibody specifically detects C2CD4C in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
C2CD4C | |
Polyclonal | |
Unconjugated | |
C2CD4C | |
C2 calcium dependent domain containing 4C; C2 calcium-dependent domain containing 4C; C2 calcium-dependent domain-containing protein 4C; C2CD4C; FAM148C; family with sequence similarity 148, member C; KIAA1957; NLF3; nuclear localized factor 3; nuclear-localized factor 3; Protein FAM148C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126567 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TF44 | |
C2CD4C | |
Recombinant protein corresponding to Human C2CD4C | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only