missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2orf56 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 28025-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The function of this protein remains unknown.Spezifikation
C2orf56 | |
Polyclonal | |
Unconjugated | |
NDUFAF7 | |
C2orf56, NDUFAF7, PRO1853 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
298748, 55471, 73694 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5XI79, Q7L592, Q9CWG8 | |
NDUFAF7 | |
C2orf56 Fusion Protein Ag27590 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |