Learn More
C3orf30 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA564024
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: QAERRTSGQI DGRLAMPSDQ RGSRQTDHRM AGQSERRASE QMDRRMSGEA ERRTSEQITH RLSKLSERRP SVQIDSGSSV PSDQSP Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 42%.
C3orf30 is a protein coding gene.Spezifikation
C3orf30 | |
Polyclonal | |
Unconjugated | |
C3orf30 | |
C3orf30; chromosome 3 open reading frame 30; Testis-specific conserved, cAMP-dependent type II PK-anchoring protein; Testis-specific expressed protein 55; TEX55; TSCPA; uncharacterized protein C3orf30 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
152405 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96M34 | |
C3orf30 | |
Recombinant protein corresponding to Human C3orf30 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only