Learn More
C4orf14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557420
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: VLGNKVDLLP QDAPGYRQRL RERLWEDCAR AGLLLAPGHQ GPQRPVKDEP QDGENPNPPN WSRTVVRDVR LISAKTGYGV Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 75%.
The protein encoded by this gene is a nuclear-encoded GTPase that functions in the mitochondrion. Upon translation, this protein is imported into the nucleus and then into the nucleolus before being exported to the mitochondrion. The encoded protein is required for oxygen-dependent regulation of mitochondrial respiratory complexes and for mitochondrial protein synthesis.Spezifikation
C4orf14 | |
Polyclonal | |
Unconjugated | |
NOA1 | |
2610024G14Rik; AtNOS1; AU022345; C4orf14; hAtNOS1; hNOA1; mAtNOS1; mitochondrial GTPase 3 homolog; MMR_HSR1 domain containing protein RGD1359460; MNCb-4931; MTG3; nitric oxide associated 1; nitric oxide associated-1; nitric oxide synthase, mitochondrial (putative); nitric oxide-associated protein 1; Noa1; putative ortholog of Arabidopsis mitochondrial nitric oxide synthase; RGD1359460 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84273 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NC60 | |
NOA1 | |
Recombinant protein corresponding to Human C4orf14 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only