missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C4orf19 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534718
Additional Details : Netto-Gewicht : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
C4orf19 is a protein coding gene. Gene ontology (GO) annotation include cell junction; nucleoplasm.Specifications
C4orf19 | |
Polyclonal | |
Unconjugated | |
C4orf19 | |
C4orf19; CD019; chromosome 4 open reading frame 19; uncharacterized protein C4orf19 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55286 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8IY42 | |
C4orf19 | |
Recombinant fragment corresponding to a region within amino acids 127 and 314 of C4orf19 (Uniprot ID#Q8IY42) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |