Learn More
C9orf75 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544400
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: PPFLPAASEE AEPAEGLRVP GLAKNSREYV RPGLPVTFID EVDSEEAPQA Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 100%
C9orf75 Polyclonal antibody specifically detects C9orf75 in Human samples. It is validated for Western Blot.Spezifikation
C9orf75 | |
Polyclonal | |
Unconjugated | |
TPRN | |
C430004E15Rik; C87750; C9orf75; DFNB79; LOW QUALITY PROTEIN: taperin; taperin; TPRN | |
Rabbit | |
Affinity Chromatography | |
RUO | |
286262 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q4KMQ1 | |
TPRN | |
Synthetic peptide directed towards the middle region of human C9orf75 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |