missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CADM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA316744
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Suggested positive control: rat brain.
Mediates homophilic cell-cell adhesion in a Ca (2+)-independent manner. Also mediates heterophilic cell-cell adhesion with CADM3 and PVRL3 in a Ca (2+)-independent manner. Acts as a tumor suppressor in non-small-cell lung cancer (NSCLC) cells. Interaction with CRTAM promotes natural killer (NK) cell cytotoxicity and interferon-gamma (IFN-gamma) secretion by CD8+ cells in vitro as well as NK cell-mediated rejection of tumors expressing CADM3 in vivo. May contribute to the less invasive phenotypes of lepidic growth tumor cells. In mast cells, may mediate attachment to and promote communication with nerves. CADM1, together with MITF, is essential for development and survival of mast cells in vivo. May act as a synaptic cell adhesion molecule that drives synapse assembly. May be involved in neuronal migration, axon growth, pathfinding, and fasciculation on the axons of differentiating neurons. May play diverse roles in the spermatogenesis including in the adhesion of spermatocytes and spermatids to Sertoli cells and for their normal differentiation into mature spermatozoa.Spezifikation
CADM1 | |
Polyclonal | |
Unconjugated | |
CADM1 | |
2900073G06Rik; 3100001I08Rik; AI987920; Bl2; cadm1; Cell adhesion molecule 1; cell adhesion molecule 1 transcript variant 8abc; Igsf4; Igsf4a; Immunoglobulin superfamily member 4; immunoglobulin superfamily member 4A; immunoglobulin superfamily, member 4; immunoglobulin superfamily, member 4A; immunoglobulin superfamily, member 4D variant 1; immunoglobulin superfamily, member 4D variant 2; immunosuperfamily protein Bl2; Necl2; NECL-2; nectin-like 2; nectin-like protein 2; RA175; RA175A; RA175B; RA175C; RA175N; secretory isoform of TSLC1; sgigsf; Spermatogenic immunoglobulin superfamily; ST17; sTSLC-1; Synaptic cell adhesion molecule; SynCAM; SynCam1; truncated CADM1 protein; TSLC1; TSLC-1; TSLC1/Nectin-like 2/IGSF4; tumor suppressor in lung cancer 1; Tumor suppressor in lung cancer 1 homolog | |
Rabbit | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
Conc. Not Determined | |
whole serum with 0.02% sodium azide | |
Q6AYP5, Q9BY67 | |
CADM1 | |
A synthetic peptide corresponding to the C-terminal region of the rat protein. | |
RUO | |
23705, 363058 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |