Learn More
Calnexin Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Marke: Proteintech 81938-1-RR-100UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Calnexin antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Calnexin. This antibody recognizes human, mouse, rat antigen. The Calnexin antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Spezifikation
| Calnexin | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| CNX; FLJ26570; IP90; P90 | |
| Recombinant Protein | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 90 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 3K18 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 821 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |