Learn More
CASD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA560700
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SIAKPHVIVA GAATWSIKIH NGSSEALSQY KMNITSIAPL LEKLAKTSDV YWVLQDPVYE DLLSENRKMI TNEKIDAY Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
O-acetyltransferase that catalyzes 9-O-acetylation of sialic acids (PubMed:20947662, PubMed:26169044). Sialic acids are sugars at the reducing end of glycoproteins and glycolipids, and are involved in various processes such as cell-cell interactions, host-pathogen recognition (PubMed:20947662, PubMed:26169044).Spezifikation
CASD1 | |
Polyclonal | |
Unconjugated | |
CASD1 | |
C7orf12; Cas1; CAS1 domain containing 1; CAS1 domain-containing protein 1; Casd1; Cast1; N-acetylneuraminate 9-O-acetyltransferase; Nbla04196; O-acetyltransferase; Sialate O-acetyltransferase; SOAT; WUGSC:H_GS542D18.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64921 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96PB1 | |
CASD1 | |
Recombinant protein corresponding to Human CASD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only