missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CASKIN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA524430
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This gene encodes a large protein that contains six ankyrin repeats, as well as a Src homology 3 (SH3) domain and two sterile alpha motif (SAM) domains, which may be involved in protein-protein interactions. The C-terminal portion of this protein is proline-rich and contains a conserved region. A related protein interacts with calcium/calmodulin-dependent serine protein kinase (CASK). Alternative splicing results in multiple transcript variants.Spezifikation
CASKIN2 | |
Polyclonal | |
Unconjugated | |
CASKIN2 | |
1600028L06Rik; ANKS5B; CASK interacting protein 2; Caskin2; Caskin-2; cask-interacting protein 2; KIAA1139; mKIAA1139 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
57513 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 0.09% sodium azide | |
Q8WXE0 | |
CASKIN2 | |
KLH conjugated synthetic peptide between 714-741 amino acids from the central region of human CASKIN2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |