Learn More
Catalase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529650
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: Molt-4, Raji, rat kidney, CAT-transfected 293T. Predicted reactivity: Mouse (93%), Rat (92%), Zebrafish (82%), Dog (95%), Pig (94%), Chicken (87%), Bovine (93%), Guinea pig (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kDa in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara's disease.Spezifikation
Catalase | |
Polyclonal | |
Unconjugated | |
Cat | |
2210418N07; Cas1; Cas-1; CAT; Cat01; catalase; catalase 1; Catl; CS1; Cs-1; CTA1; HGNC:1516; hypothetical protein LOC531682; MGC138422; MGC138424 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
24248, 847 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P04040, P04762 | |
Cat | |
Recombinant fragment corresponding to a region within amino acids 89 and 341 of Human Catalase | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |