Learn More
Cathepsin B Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA547975
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, approximately 25% cross-reactivity with recombinant human Cathepsin B is observed and less than 5% cross-reactivity with recombinant mouse (rm) Cathepsin A, rmCathepsin C, and rmCathepsin D is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Cathepsin B Polyclonal antibody specifically detects Cathepsin B in Mouse samples. It is validated for Immunohistochemistry (Frozen), Neutralization, Western Blot.Spezifikation
Cathepsin B | |
Polyclonal | |
Unconjugated | |
Ctsb | |
amyloid precursor protein secretase; APP secretase; APPS; cathepsin B; Cathepsin B heavy chain; Cathepsin B light chain; Cathepsin B1; CB; CPSB; Ctsb; cysteine protease; RSG-2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
13030 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Neutralization, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P10605 | |
Ctsb | |
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin B His18-Phe339 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |