missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CCBL1 (Human) Recombinant Protein
Human CCBL1 full-length ORF ( AAH33685, 1 a.a. - 374 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00000883-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
- Sequence: MAKQLQARRLDGIDYNPWVEFVKLASEHDVVNLGQGFPDFPPPDFAVEAFQHAVSGDFMLNQYTKTFVIIIEPFFDCYEPMTMMAGGRPVFVSLKPGPIQNGELGSSSNWQLDPMELAGKFTSRTKALVLNTPNNPLGKVFSREELELVASLCQQHDVVCITDEVYQWMVYDGHQHISIASLPGMWERTLTIGSAGKTFSATGWKVGWVLGPDHIMKHLRTVHQNSVFHCPTQSQAAVAESFEREQLLFRQPSSYFVQFPQAMQRCRDHMIRSLQSVGLKPIIPQGSYFLITDISDFKRKMPDLPGAVDEPYDRRFVKWMIKNKGLVAIPVSIFYSVPHQKHFDHYIRFCFVKDEATLQAMDEKLRKWKVELWP
Spezifikation
AAH33685 | |
cysteine conjugate-beta lyase, cytoplasmic | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CCBL1 | |
GST |
883 | |
Wheat germ expression system | |
25 μg | |
FLJ95217, GTK, KAT1, KATI, MGC29624 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |