missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCL24/Eotaxin 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 22306-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Eotaxin 2 belongs to the subfamily of small cytokine CC genes. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene displays chemotactic activity on resting T lymphocytes, a minimal activity on neutrophils, and is negative on monocytes and activated T lymphocytes. The protein is also a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell line.Spezifikation
| CCL24/Eotaxin 2 | |
| Polyclonal | |
| Unconjugated | |
| CCL24 | |
| C C motif chemokine 24, CCL24, CK beta 6, Ckb 6, Eotaxin 2, MPIF 2, MPIF2, SCYA24, Small inducible cytokine A24 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56221, 6369 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00175, Q9JKC0 | |
| CCL24 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur