missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR7 (CD197) Rat anti-Human, BB700, Clone: 3D12, BD Horizon™
Rat Monoclonal Antibody
Marke: BD Biosciences 566437
Weitere Details : Netto-Gewicht : 0.00760kg
Beschreibung
CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow Cytometry.Spezifikation
CCR7 (CD197) | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
100 Tests | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |
Flow Cytometry | |
3D12 | |
Rat | |
RUO | |
Human | |
Purified |