missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD106 Mouse anti-Human, Oligo, Clone: 51-10C9, BD™ AbSeq ™
Mouse Monoclonal Antibody
Marke: BD Biosciences 940375
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CD106 Monoclonal antibody specifically detects CD106 in Human samples. It is validated for Single Cell 3' SequencingSpezifikation
CD106 | |
Monoclonal | |
Oligo | |
VCAM-1; Vascular cell adhesion protein 1; INCAM-100; L1CAM | |
Human VCAM-1 Recombinant Protein | |
RUO | |
Human | |
IgG1 κ |
Single Cell 3' Sequencing | |
Clone 51-10C9 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |