missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD117/c-kit Antibody (9D2) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Marke: Novus Biologicals NB110-93600-0.025MG
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD117/c-kit Monoclonal specifically detects CD117/c-kit in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Spezifikation
| CD117/c-kit | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:100-1:1000, Immunohistochemistry 1:100-1:400, Immunohistochemistry-Paraffin 1:100-1:400 | |
| P05532 | |
| KIT | |
| Recombinant Mouse SCF-R extracellular domain | |
| 0.025 mg | |
| Cancer, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases | |
| 3815 | |
| Mouse | |
| Purified |
| Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry | |
| 9D2 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction