Learn More
CD117/c-kit Antibody (9D2) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
201.00 € - 518.00 €
Spezifikation
| Antigen | CD117/c-kit |
|---|---|
| Klon | 9D2 |
| Konzentration | LYOPH |
| Verdünnung | Western Blot 1:100-1:1000, Immunohistochemistry 1:100-1:400, Immunohistochemistry-Paraffin 1:100-1:400 |
| Anwendungen | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry |
Beschreibung
CD117/c-kit Monoclonal specifically detects CD117/c-kit in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Spezifikation
| CD117/c-kit | |
| LYOPH | |
| Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Cancer, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases | |
| P05532 | |
| 3815 | |
| IgG2 | |
| Protein G purified |
| 9D2 | |
| Western Blot 1:100-1:1000, Immunohistochemistry 1:100-1:400, Immunohistochemistry-Paraffin 1:100-1:400 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein | |
| KIT | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |