missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c Mouse anti-Human, Oligo, Clone: 3.9, BD™ AbSeq ™
Mouse Monoclonal Antibody
Marke: BD Biosciences 940363
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CD11c Monoclonal antibody specifically detects CD11c in Human samples. It is validated for Single Cell 3' SequencingSpezifikation
CD11c | |
Monoclonal | |
Oligo | |
ITGAX; AlphaX Integrin; Axb2; Integrin alpha-X; CR4; SLEB6; p150,95 alpha | |
Human monocytes and synovial cells | |
RUO | |
Human | |
IgG1 κ |
Single Cell 3' Sequencing | |
Clone 3.9 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |