missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD138 (Syndecan-1) Mouse, anti-Human, APC, Clone: MI15, STEMCELL Technologies
Mouse Monoclonal Antibody
237.60 € - 607.50 €
Spezifikation
| Antigen | CD138 (Syndecan-1) |
|---|---|
| Klon | MI15 |
| Anwendungen | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Klassifikation | Monoclonal |
| Konjugat | APC |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
17118341
|
Stemcell Technologies
60003AZ.1 |
25 Tests |
237.60 €
25 Tests |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
17108341
|
Stemcell Technologies
60003AZ |
100 Tests |
607.50 €
100 Tests |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
- The MI15 antibody reacts with an extracellular epitope on CD138 (Syndecan-1), an ∼85 - 95 kDa type 1 transmembrane glycoprotein expressed on the surface of pre-B cells, immature B cells, and normal and malignant plasma cells (but not mature circulating B cells), as well as on non-hematopoietic cells such as embryonic mesenchymal cells, endothelial, epithelial and neural cells
- CD138 expression is used as a diagnostic marker for several types of tumors
- CD138 is thought to act primarily as a receptor that modulates cell proliferation, cell migration and cell-matrix associations by linking the extracellular matrix to the cytoskeleton
- Theparin sulfate and chondroitin sulfate moieties attached to CD138 associate with several proteins, including collagens, fibronectin, tenascin, thrombospondin and certain cytokines
- The MI15 antibody recognizes a different epitope than that of the clone DL-101 anti-CD138 antibody but blocks binding of clone B-B4
Spezifikation
| CD138 (Syndecan-1) | |
| Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| APC | |
| Cynomolgus, Human, Rhesus Monkey | |
| Primary |
| MI15 | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.
Name des Produkts