Learn More
CD14 Mouse, anti-Human, FITC, Clone: MoP9, STEMCELL Technologies
Mouse Monoclonal Antibody
Marke:  Stemcell Technologies 60124FI
Encompass_Preferred
Artikelnummer. 17179151
  • 1344.60 € / 100 Tests
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
  • The MoP9 antibody reacts with CD14, a 53 - 55 kDa glycosylphosphatidylinositol (GPI)-anchored transmembrane glycoprotein expressed at high levels on the surface of peripheral blood monocytes and macrophages, and at lower levels on granulocytes
  • An approximate 10-fold difference in expression levels between monocytes/macrophages and granulocytes makes CD14 a useful marker for distinguishing these cell populations
  • CD14 is also found on tissue macrophages, Langerhans cells, and dendritic cells
  • CD14 functions as a high-affinity receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein and modulates LPS-dependent signal transduction during the immune response to gram-negative pathogens by acting as a co-receptor for TLR 4 and MD-2
  • This triggers activation of NF-κB, cytokine secretion, and induction of the inflammatory response
  • Two soluble forms of CD14 have also been described (∼48 and ∼55 kDa)
Spezifikation
Spezifikation
CD14
Monoclonal
FITC
100 Tests
Human
Monocytes
Flow Cytometry
MoP9
Mouse
Primary
IgG2b κ
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen