Learn More
CD14 Mouse anti-Human, Unconjugated, Clone: UCHM-1, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 65056-1-IG-100UG
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CD14 is a 50-55 kDa glycosylphosphatidylinositol-anchored glycoprotein preferentially expressed on monocytes and macrophages, and at lower levels on granulocytes (PMID: 3385210; 2462937; 7685797). CD14 can also exist as a soluble protein. CD14 acts as a co-receptor for bacterial liposaccharides (LPS) (PMID: 1698311). It plays a major role in the inflammatory response of monocytes to LPS.
Spezifikation
CD14 | |
Monoclonal | |
0.1 mg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig | |
IgG2a |
Flow Cytometry | |
UCHM-1 | |
Unconjugated | |
100 μg | |
929 | |
Liquid |