Learn More
CD1a Mouse anti-Human, PE, Clone: HI149, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 555807
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
The HI149 monoclonal antibody specifically binds to CD1a. CD1a is a type I transmembrane glycoprotein. The 49 kDa CD1a polypeptide is associated with β2-microglobulin. CD1a is expressed on cortical thymocytes, dendritic cells and Langerhans cells. CD1a has structural similarities to the MHC class I antigen, and plays a role in antigen presentation.
Spezifikation
CD1a | |
Monoclonal | |
PE | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
HI149 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.