Learn More
CD200 Receptor Monoclonal Antibody (OX108), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
2985.00 € - 14910.00 €
Spezifikation
| Antigen | CD200 Receptor |
|---|---|
| Klon | OX108 |
| Konzentration | 1 mg/mL |
| Anwendungen | Flow Cytometry, Functional Assay |
| Klassifikation | Monoclonal |
Beschreibung
The MAb OX108 recognizes human CD200R also known as OX2. CD200R is an inhibitory receptor with a similar structure as its ligand, CD200, yet lacks an ITIM domain typically found in inhibitory receptors. Instead the CD200R cytoplasmic domain contains a novel phosphotyrosine binding domain, NPXY, which after binding SHIP inhibits ERK, JNK and MAPK p38 pathways. In activated macrophages signaling results in inhibition of TNFalpha secretion. Isoforms of CD200R have been identified and are thought to play a major role in differentiation, especially in regards to tolerogenic DCs. Expression is restricted to hematopoietic cells: myeloid cells (monocytes, macrophages, DCs, neutrophils, mast cells and basophils) and a subset of T lymphocytes as well as langerhans (LC) cells and dendritic epidermal T cells. The epitope of OX108 is thought to be near the binding site of CD200.
CD200 Receptor Monoclonal antibody specifically detects CD200 Receptor in Human samples. It is validated for Flow Cytometry, Functional AssaySpezifikation
| CD200 Receptor | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| antigen identified by monoclonal antibody MRC OX-2 receptor; CD200 cell surface glycoprotein receptor; CD200 R1; CD200 receptor 1; CD200R; CD200R1; cell surface glycoprotein CD200 receptor 1; Cell surface glycoprotein OX2 receptor 1; cell surface glycoprotein receptor CD200; CRTR2; HCRTR2; MOX2 receptor; Mox2r; Orexin receptor 2; OX102 antigen; OX2 receptor; Ox2r; UNQ2522/PRO6015 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| OX108 | |
| Flow Cytometry, Functional Assay | |
| Functional Grade | |
| Mouse | |
| Human | |
| Q8TD46 | |
| 131450 | |
| Primary | |
| 4° C | |
| CD200R1 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.