Learn More
Invitrogen™ CD22 Recombinant Rabbit Monoclonal Antibody (3F2M10)

Rabbit Recombinant Monoclonal Antibody
176.00 € - 436.00 €
Spezifikation
| Antigen | CD22 |
|---|---|
| Klon | 3F2M10 |
| Konzentration | 1.34 mg/mL |
| Inhalt und Lagerung | -20°C, Avoid Freeze/Thaw Cycles |
| Anwendungen | ELISA, Western Blot |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
30283763
|
Invitrogen™
MA558067 |
20 μL |
176.00 €
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
30283398
![]() |
Invitrogen™
MA558068 |
100 μL |
436.00 €
100 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Immunogen sequence: DSSKWVFEHP ETLYAWEGA DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSKWVFEHP DSSK...
CD22 (BL-CAM) is a type 1 integral membrane glycoprotein with molecular weight of 130 to 140 kDa. CD22 is expressed in both the cytoplasm and cell membrane of B-lymphocytes. CD22 antigen appears early in B-cell lymphocyte differentiation at approximately the same stage as the CD19 antigen. Unlike other B-cell markers, CD22 membrane expression is limited to the late differentiation stages comprised between mature B cells (CD22+) and plasma cells (CD22-), and may thus prove useful in phenotyping mature leukemias. CD22 is also strongly expressed in hairy cell leukemia. CD22 preferentially binds to alpha2,6-linked sialic acid. The sialic acid recognition site can be masked by cis interactions with sialic acids on the same cell surface. Upon ligand induced tyrosine phosphorylation in the immune response seems to be involved in regulation of B cell antigen receptor signaling. CD22 plays a role in positive regulation through interaction with Src family tyrosine kinases and may also act as an inhibitory receptor by recruiting cytoplasmic phosphatases via their SH2 domains that block signal transduction through dephosphorylation of signaling molecules. CD22 is also strongly expressed in hairy cell leukemia. CD22 is also positive in diffuse large B-cell lymphoma and nodular lymphocyte predominance Hodgkin's lymphoma, but negative in classical Hodgkin's lymphoma.Spezifikation
| CD22 | |
| 1.34 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P20273 | |
| 933 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-687 of human CD22 (NP_001762.2). | |
| Antibody |
| 3F2M10 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| CD22 | |
| A530093D23; B-cell receptor CD22; BL-CAM; B-lymphocyte cell adhesion molecule; B-lymphocyte cell adhesion molecule (BL-CAM); Cd22; CD22 antigen; CD22 molecule; FLJ22814; Lectin 2; Leu-14; Lyb8; Lyb-8; MGC130020; sialic acid binding Ig-like lectin 2; sialic acid-binding Ig-like lectin 2; Sialic acid-binding Ig-like lectin 2 (Siglec-2); Siglec2; Siglec-2; T-cell surface antigen Leu-14 | |
| CD22 | |
| Primary | |
| Protein A |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.