missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD223 (LAG-3) Mouse anti-Human, BUV615, Clone: T47-530, BD OptiBuild™
Mouse Monoclonal Antibody
Marke: BD Biosciences 752362
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
CD223 (LAG-3) Monoclonal antibody specifically detects CD223 (LAG-3) in Human samples. It is validated for Flow Cytometry.
Spezifikation
CD223 (LAG-3) | |
Monoclonal | |
0.2 mg/mL | |
Hu LAG-3 (CD223) BUV615 T47-530 50 μg | |
Human LAG-3 Recombinant Protein | |
RUO | |
CD223 (LAG-3) | |
Refrigerate (2 to 8°C) | |
Human |
Flow Cytometry | |
T47-530 | |
Brilliant Ultraviolet 615 | |
Mouse | |
1 x 50 μg | |
Primary | |
Human | |
IgG1 κ |