Learn More
CD235a (Glycophorin A) Monoclonal Antibody (HIR2 (GA-R2)), Super Bright™ 702, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
161.00 € - 350.00 €
Spezifikation
Antigen | CD235a (Glycophorin A) |
---|---|
Klon | HIR2 (GA-R2) |
Konzentration | 5 μL/Test |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15933794
|
Affymetrix eBioscience
67-9987-42 |
100 Tests |
350.00 €
100 Stück |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen | Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15923794
|
Affymetrix eBioscience
67-9987-41 |
25 Tests |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Description: The 17A2 monoclonal antibody reacts with the mouse CD3 complex. CD3 subunits gamma, delta and epsilon are required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Binding of 17A2 to CD3 initiates the intracellular biochemical pathway resulting in cellular activation and proliferation. Applications Reported: This 17A2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 17A2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 506 can be excited with the violet laser line (405 nm) and emits at 506 nm. We recommend using a 510/20 band pass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. Excitation: 405 nm; Emission: 506 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD235a (Glycophorin A) Monoclonal antibody specifically detects CD235a (Glycophorin A) in Human samples. It is validated for Flow Cytometry.Spezifikation
CD235a (Glycophorin A) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P02724 | |
GYPA | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
GYPA |
HIR2 (GA-R2) | |
Flow Cytometry | |
Super Bright 702 | |
Mouse | |
Human | |
2993 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |