missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25 Monoclonal Antibody (PC61.5), Alexa Fluor™ 532, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Marke: Affymetrix eBioscience 58-0251-80
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Rat Monoclonal Antibody
CD25 Monoclonal antibody specifically detects CD25 in Mouse samples. It is validated for Flow CytometrySpezifikation
CD25 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
P01590 | |
IL2RA | |
Affinity chromatography | |
RUO | |
16184 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
PC61.5 | |
Alexa Fluor 532 | |
IL2RA | |
BOS_12822; CD25; IDDM10; IL 2 RA; IL 2 receptor; IL 2R; IL 2R subunit alpha; IL2 RA; IL2 RA antibody; IL2 receptor; IL2 receptor subunit alpha; IL-2 receptor subunit alpha; Il2r; IL2R subunit alpha; Il2ra; IL-2RA; Il2ra antibody; IL2RAC; IMD41; Interleukin2 receptor subunit alpha; Interleukin-2 receptor subunit alpha; Ly-43; p55; sIL 2R; soluble IL 2 receptor; TAC antigen; TCGFR | |
Rat | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 λ |