Learn More
CD3 Mouse, anti-Human, FITC, Clone: SK7, STEMCELL Technologies
Mouse Monoclonal Antibody
Marke: Stemcell Technologies 60127FI.1
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- The SK7 antibody reacts with the ∼20 kDa CD3ε subunit of the human T cell receptor (TCR)/CD3 complex, which is expressed on the surface of ∼95% of mature T cells and NKT cells, and variably on thymocytes
- A majority of T cell neoplasms also express CD3
- The CD3 complex, which is assembled from combinations of CD3γ, δ, ε, η and ζ subunits, associates non-covalently with the TCR and is involved in transducing antigen recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR
- Activation of T cells by the TCR involves the cytoplasmic tails of the CD3 subunits, which are structurally related type 1 transmembrane proteins and members of the immunoglobulin superfamily
- Mutations in the CD3 subunits have been associated with various immunodeficiency disorders, including severe combined immunodeficiency (SCID)
Spezifikation
| CD3 | |
| Monoclonal | |
| FITC | |
| 25 Tests | |
| Chimpanzee, Human | |
| T Cells |
| Flow Cytometry | |
| SK7 | |
| Mouse | |
| Primary | |
| IgG1 κ |
Berichtigung von Produktinhalten
Das Fisher Scientific Encompass-Programm bietet Artikel an, die nicht Teil unseres Vertriebsportfolios sind. Diese Produkte verfügen in der Regel nicht über Bilder oder detaillierte Beschreibungen. Wir sind jedoch bestrebt, Ihr Einkaufserlebnis zu verbessern. Bitte verwenden Sie das untenstehende Formular, um uns Feedback zum Inhalt dieses Produkts zu geben.