missing translation for 'onlineSavingsMsg'
Learn More

CD318 (CDCP1) Monoclonal Antibody (CSTEM26), eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Affymetrix eBioscience 14-3189-82

166.67 EUR gültig bis 2024-03-29
Benutzen Sie den Promotions-Code "21615" um Ihren Promotions-Preis zu erhalten.



Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15885343

  • 250.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This monoclonal antibody CSTEM26 reacts with human CD318, also known as CUB domain-containing protein 1 (CDCP1) and Trask. Applications Reported: This CSTEM26 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CSTEM26 antibody has been tested by flow cytometric analysis of normal human induced pluripotent stem cells. This may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

CDCP1 is a transmembrane protein containing three extracellular CUB domains. It is found to be overexpressed in colon and lung cancers. Its expression level is correlated with the metastatic ability of carcinoma cells. CDCP1 is located on the cell surface. It has been shown to be tyrosine phosphorylated in a cancer cell line. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Spezifikation

Spezifikation

CD318 (CDCP1)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q9H5V8
Cdcp1
Affinity chromatography
RUO
64866
4° C, do not freeze
Liquid
Flow Cytometry
CSTEM26
Unconjugated
Cdcp1
9030022E12Rik; AA409659; CD318; CDCP1; CUB domain containing protein 1; CUB domain-containing protein 1; E030027H19Rik; gp140; Membrane glycoprotein gp140; RGD1305578; SIMA135; Subtractive immunization M plus HEp3-associated 135 kDa protein; transmembrane and associated with src kinases; TRASK; UNQ2486/PRO5773
Mouse
100 μg
Primary
Human
Antibody
IgG2a κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote