missing translation for 'onlineSavingsMsg'
Learn More

CD45 Mouse anti-Human, PE-Cy7, Clone: HI30, BD

Mouse Monoclonal Antibody

221.00 € - 560.00 €

Spezifikation

Antigen CD45
Klon HI30
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Konjugat PE-Cyanine7
View More Specs

Products 2
Product Code Brand Menge Price Quantity & Availability  
Product Code Brand Menge Price Quantity & Availability  
15890589
missing translation for 'viewDocuments' Promotion Details
BD Biosciences
560915
25 Tests
221.00 €
1 Stück
View alternative products
missing translation for 'toSaveUpto' 
»
Verfügbar ab: 07-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
15879028
missing translation for 'viewDocuments' Promotion Details
BD Biosciences
557748
100 Tests
560.00 €
1 Stück
View alternative products
missing translation for 'toSaveUpto' 
»
Verfügbar ab: 31-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Description

Description

The HI30 monoclonal antibody specifically binds to the 180, 190, 205, 220 kDa protein isoforms of CD45. CD45 is encoded by the PTPRC (Protein tyrosine phosphatase receptor type C) gene. CD45, also known as the leukocyte common antigen (LCA), is a member of the protein tyrosine phosphatase (PTP) family. It is present on all human leukocytes including lymphocytes, monocytes, granulocytes, eosinophils, and thymocytes. CD45 is absent from circulating erythrocytes, platelets, or mature erythroid cells of bone marrow and non-hemopoietic tissues.

Specifications

Specifications

CD45
Flow Cytometry
PE-Cyanine7
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
PTPRC; CD45R; LCA; L-CA; Leukocyte Common Antigen; B220; T200; GP180; LY5
HI30
Monoclonal
Mouse
Human
Human Peripheral Blood Leucocytes
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Certificates

For Research Use Only.