Learn More
CD45R Mouse anti-Rat, FITC, Clone: HIS24, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 554880
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The HIS24 antibody reacts with a developmentally regulated form of CD45 found on most B lymphocytes, including developing B cells in the bone marrow and peripheral B cells, but not plasma cells. The level of expression of this CD45R antigen appears to be an indicator of both maturational stages in the B-cell lineage and of functionally distinct B-cell subsets. The HIS24 mAb, in combination with other markers such as TdT and Ig expression, is effective in the identification of B-cell progenitors. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the rat are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Spezifikation
CD45R | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Low-Density Cells from AO/G Rat Peyer's Patches | |
0.5 mg | |
Immunology | |
Rat | |
IgG2b κ |
Flow Cytometry | |
HIS24 | |
FITC | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.